3-Bromodibenzofuran 97%
|
|
BLDPBD247299-250MGEA
7.3
EUR
BLDPBD247299-250MG
BLDPBD247299-25G
BLDPBD247299-10G
BLDPBD247299-1G
BLDPBD247299-100G
BLDPBD247299-5G
3-Bromodibenzofuran 97%
3-Bromodibenzofuran
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Kaava:
C₁₂H₇BrO MW: 247,09 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD09966042 CAS-numero: 26608-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...