3-Bromo-4-nitro-1H-pyrazole 96%

BLDPBD247349-100MGEA 75.9 EUR
BLDPBD247349-100MG BLDPBD247349-5G BLDPBD247349-1G BLDPBD247349-250MG
3-Bromo-4-nitro-1H-pyrazole 96%
3-Bromo-4-nitro-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₃H₂BrN₃O₂
MW: 191,971 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD19441764
CAS-numero: 784193-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR