2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene 97%
|
|
BLDPBD247418-100MGEA
39.5
EUR
BLDPBD247418-100MG
BLDPBD247418-1G
BLDPBD247418-250MG
2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene 97%
2,6-Dibromo-4,4-bis(2-ethylhexyl)-4H-cyclopenta[2,1-b:3,4-b']dithiophene
Storage condition: Inert atmosphere, 2-8°C
Kaava:
C₂₅H₃₆Br₂S₂ MW: 560,49 g/mol Storage Temperature: Jääkaappi |
MDL numero:
MFCD16619302 CAS-numero: 365547-21-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...