4,6-Dichloropyrimidin-2-ol 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD248834-100MGEA 108 EUR
BLDPBD248834-100MG BLDPBD248834-1G BLDPBD248834-250MG
4,6-Dichloropyrimidin-2-ol 95%
4,6-Dichloropyrimidin-2-ol

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₄H₂Cl₂N₂O
MW: 164,977 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09033880
CAS-numero: 6297-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR