2-Amino-3-methoxypyridine 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD25082-5GEA 40.6 EUR
BLDPBD25082-5G BLDPBD25082-10G BLDPBD25082-100G BLDPBD25082-1G BLDPBD25082-25G
2-Amino-3-methoxypyridine 98%
2-Amino-3-methoxypyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₆H₈N₂O
MW: 124,1400 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD07374874
CAS-numero: 10201-71-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR