1,4-Diamino-2,3-dichloro-9,10-anthraquinone 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD252026-5GEA 13.6 EUR
BLDPBD252026-5G BLDPBD252026-25G BLDPBD252026-100G
1,4-Diamino-2,3-dichloro-9,10-anthraquinone 95%
1,4-Diamino-2,3-dichloro-9,10-anthraquinone

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₄H₈Cl₂N₂O₂
MW: 307,1315 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00035693
CAS-numero: 81-42-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR