2-Bromo-4-methyl-1-nitrobenzene 98%

BLDPBD254198-1GEA 35.4 EUR
BLDPBD254198-1G BLDPBD254198-10G BLDPBD254198-5G BLDPBD254198-25G BLDPBD254198-250MG
2-Bromo-4-methyl-1-nitrobenzene 98%
2-Bromo-4-methyl-1-nitrobenzene

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₆BrNO₂
MW: 216,03 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07363742
CAS-numero: 40385-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR