4,7-Dichloro-1,10-phenanthroline 97%

BLDPBD254685-100MGEA 7.3 EUR
BLDPBD254685-100MG BLDPBD254685-5G BLDPBD254685-25G BLDPBD254685-250MG BLDPBD254685-1G
4,7-Dichloro-1,10-phenanthroline 97%
4,7-Dichloro-1,10-phenanthroline

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₂H₆Cl₂N₂
MW: 249,09 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00233882
CAS-numero: 5394-23-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR