5-Bromo-m-cresol 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD25521-250MGEA 10.4 EUR
BLDPBD25521-250MG BLDPBD25521-5G BLDPBD25521-25G BLDPBD25521-10G BLDPBD25521-1G
5-Bromo-m-cresol 98%
5-Bromo-m-cresol

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₇BrO
MW: 187,0300 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09038281
CAS-numero: 74204-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR