3-Bromo-5-methoxyquinoline 98+%

BLDPBD256547-1GEA 1150 EUR
BLDPBD256547-1G BLDPBD256547-250MG BLDPBD256547-100MG
3-Bromo-5-methoxyquinoline 98+%
3-Bromo-5-methoxyquinoline

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₀H₈BrNO
MW: 238,0800 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD22690582
CAS-numero: 1416438-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR