2-Chloro-4-hydroxybenzoic acid hydrate 98%

BLDPBD257513-5GEA 80 EUR
BLDPBD257513-5G BLDPBD257513-1G BLDPBD257513-250MG
2-Chloro-4-hydroxybenzoic acid hydrate 98%
2-Chloro-4-hydroxybenzoic acid hydrate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₅ClO₃·xH₂O
MW: 172,57 g/mol (anhy)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00798116
CAS-numero: 440123-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR