7-Methoxy-2-methyl-1H-indole 95%

BLDPBD258243-250MGEA 223 EUR
BLDPBD258243-250MG BLDPBD258243-50MG BLDPBD258243-100MG
7-Methoxy-2-methyl-1H-indole 95%
7-Methoxy-2-methyl-1H-indole
Kaava: C₁₀H₁₁NO
MW: 161,2 g/mol
MDL numero: MFCD12828149
CAS-numero: 53512-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR