6-Chloro-2-methylpyrimidin-4-amine 97%

BLDPBD25830-500GEA 1050 EUR
BLDPBD25830-500G BLDPBD25830-5G BLDPBD25830-1G BLDPBD25830-10G BLDPBD25830-100G BLDPBD25830-25G
6-Chloro-2-methylpyrimidin-4-amine 97%
6-Chloro-2-methylpyrimidin-4-amine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₅H₆ClN₃
MW: 143,58 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD07782029
CAS-numero: 1749-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR