1,4-Dibromo-3,5-difluorobenzene 98%

BLDPBD260769-5GEA 47.8 EUR
BLDPBD260769-5G BLDPBD260769-100G BLDPBD260769-1G BLDPBD260769-500G BLDPBD260769-25G
1,4-Dibromo-3,5-difluorobenzene 98%
1,4-Dibromo-3,5-difluorobenzene

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₆H₂Br₂F₂
MW: 271,8849 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07783665
CAS-numero: 128259-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR