6-Hydroxypyrimidine-4-carboxylic acid 97%

BLDPBD2613-250MGEA 7.3 EUR
BLDPBD2613-250MG BLDPBD2613-25G BLDPBD2613-5G BLDPBD2613-100MG BLDPBD2613-1G BLDPBD2613-100G BLDPBD2613-10G
6-Hydroxypyrimidine-4-carboxylic acid 97%
6-Hydroxypyrimidine-4-carboxylic acid

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₅H₄N₂O₃
MW: 140,0969 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00023267
CAS-numero: 6299-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR