4-Hexylthiophene-2-carbonitrile 95+%

BLDPBD262601-250MGEA 154 EUR
BLDPBD262601-250MG BLDPBD262601-100MG BLDPBD262601-1G
4-Hexylthiophene-2-carbonitrile 95+%
4-Hexylthiophene-2-carbonitrile

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₁H₁₅NS
MW: 193,31 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD18413940
CAS-numero: 1224430-39-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR