2-Cyclopropylbenzeneboronic acid 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD263124-1GEA 229 EUR
BLDPBD263124-1G BLDPBD263124-250MG BLDPBD263124-100MG
2-Cyclopropylbenzeneboronic acid 95%
2-Cyclopropylbenzeneboronic acid

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₉H₁₁BO₂
MW: 161,993 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD12760762
CAS-numero: 1373393-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR