6-(Trifluoromethyl)-1H-indazole 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD26387-5GEA 433 EUR
BLDPBD26387-5G BLDPBD26387-1G BLDPBD26387-100MG BLDPBD26387-250MG
6-(Trifluoromethyl)-1H-indazole 97%
6-(Trifluoromethyl)-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₅F₃N₂
MW: 186,1300 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD08690097
CAS-numero: 954239-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR