3-Bromodibenzothiophene 98%
|
|
BLDPBD265959-250MGEA
19.8
EUR
BLDPBD265959-250MG
BLDPBD265959-25G
BLDPBD265959-10G
BLDPBD265959-5G
BLDPBD265959-1G
3-Bromodibenzothiophene 98%
3-Bromodibenzothiophene
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Kaava:
C₁₂H₇BrS MW: 263,15 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD11052997 CAS-numero: 97511-04-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...