Dibenzothiophene-2-carboxaldehyde 98%

BLDPBD267664-100MGEA 21.9 EUR
BLDPBD267664-100MG
Dibenzothiophene-2-carboxaldehyde 98%
Dibenzothiophene-2-carboxaldehyde

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₁₃H₈OS
MW: 212,27 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD02251670
CAS-numero: 22099-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR