Ethyl-1H-indazole-4-carboxylate 95%

BLDPBD26785-250MGEA 28.1 EUR
BLDPBD26785-250MG BLDPBD26785-1G BLDPBD26785-5G
Ethyl-1H-indazole-4-carboxylate 95%
Ethyl-1H-indazole-4-carboxylate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₁₀H₁₀N₂O₂
MW: 190,20 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD07371611
CAS-numero: 885279-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR