2-(4-aminophenoxy)-2-methylpropanoic acid 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD27261-5GEA 318 EUR
BLDPBD27261-5G BLDPBD27261-100MG BLDPBD27261-250MG BLDPBD27261-1G
2-(4-aminophenoxy)-2-methylpropanoic acid 95%
2-(4-aminophenoxy)-2-methylpropanoic acid

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₁₀H₁₃NO₃
MW: 195,215 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00084883
CAS-numero: 117011-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR