Imidazo[1,2-a]pyridine-8-carboxylic acid 97%

BLDPBD27447-1GEA 25 EUR
BLDPBD27447-1G BLDPBD27447-10G BLDPBD27447-5G BLDPBD27447-25G BLDPBD27447-250MG
Imidazo[1,2-a]pyridine-8-carboxylic acid 97%
Imidazo[1,2-a]pyridine-8-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₆N₂O₂
MW: 162,1454 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD06739251
CAS-numero: 133427-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR