1-Boc-4-(4-aminophenoxy)piperidine 98%

BLDPBD27491-5GEA 337 EUR
BLDPBD27491-5G BLDPBD27491-250MG BLDPBD27491-1G
1-Boc-4-(4-aminophenoxy)piperidine 98%
1-Boc-4-(4-aminophenoxy)piperidine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: C₁₆H₂₄N₂O₃
MW: 292,3734 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04115023
CAS-numero: 138227-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR