5-Bromo-7-methyl-1H-indazole 97%

BLDPBD27641-10GEA 164 EUR
BLDPBD27641-10G BLDPBD27641-25G BLDPBD27641-100G BLDPBD27641-5G BLDPBD27641-250MG BLDPBD27641-1G
5-Bromo-7-methyl-1H-indazole 97%
5-Bromo-7-methyl-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₈H₇BrN₂
MW: 211,0600 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03990484
CAS-numero: 156454-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR