9,9-Diphenylfluorene-2-boronic acid 98%
|
|
BLDPBD279869-1GEA
51
EUR
BLDPBD279869-1G
BLDPBD279869-5G
BLDPBD279869-10G
BLDPBD279869-250MG
9,9-Diphenylfluorene-2-boronic acid 98%
9,9-Diphenylfluorene-2-boronic acid
Storage condition: Inert atmosphere, Room Temperature
Kaava:
C₂₅H₁₉BO₂ MW: 362,24 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD22581299 CAS-numero: 400607-31-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...