2-Amino-4-methyloxazole 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD28478-1GEA 58.2 EUR
BLDPBD28478-1G BLDPBD28478-5G
2-Amino-4-methyloxazole 95%
2-Amino-4-methyloxazole

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Kaava: C₄H₆N₂O
MW: 98,1032 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00126682
CAS-numero: 35629-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR