5-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD287104-1GEA 677 EUR
BLDPBD287104-1G BLDPBD287104-100MG BLDPBD287104-250MG
5-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine 95%
5-Chloro-3-iodo-1H-pyrazolo[3,4-c]pyridine
Kaava: C₆H₃ClIN₃
MW: 279,466 g/mol
MDL numero: MFCD18250844
CAS-numero: 1260666-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR