10-Acetylphenothiazine 95%

BLDPBD287893-250MGEA 9.4 EUR
BLDPBD287893-250MG BLDPBD287893-5G BLDPBD287893-100MG BLDPBD287893-1G
10-Acetylphenothiazine 95%
10-Acetylphenothiazine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₄H₁₁NOS
MW: 241,31 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00022264
CAS-numero: 1628-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR