Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide 95+%

BLDPBD289650-1GEA 183 EUR
BLDPBD289650-1G BLDPBD289650-100MG BLDPBD289650-5G BLDPBD289650-250MG
Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide 95+%
Lithium 4-methyl-1-(pyridin-2-yl)-2,6,7-trioxa-1-borabicyclo[2.2.2]octan-1-uide

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₀H₁₃BNO₃·Li
MW: 212,967 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD16038156
CAS-numero: 1014717-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR