7-Bromo-1H-indazole 97%

BLDPBD28982-250MGEA 7.3 EUR
BLDPBD28982-250MG BLDPBD28982-100G BLDPBD28982-25G BLDPBD28982-0.025G BLDPBD28982-5G BLDPBD28982-10G BLDPBD28982-1G
7-Bromo-1H-indazole 97%
7-Bromo-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Kaava: C₇H₅BrN₂
MW: 197,0300 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD07371564
CAS-numero: 53857-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR