Ethyl 2-hydroxyimidazo[1,2-a]pyridine-3-carboxylate 95%

BLDPBD289967-100MGEA 132 EUR
BLDPBD289967-100MG BLDPBD289967-1G BLDPBD289967-250MG
Ethyl 2-hydroxyimidazo[1,2-a]pyridine-3-carboxylate 95%
Ethyl 2-hydroxyimidazo[1,2-a]pyridine-3-carboxylate

Storage condition: Sealed in dry, 2-8°C

Kaava: C₁₀H₁₀N₂O₃
MW: 206,198 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD26404011
CAS-numero: 27124-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR