5-Bromo-1-methyl-4-nitro-1H-pyrazole 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD290069-100MGEA 42.6 EUR
BLDPBD290069-100MG BLDPBD290069-5G BLDPBD290069-250MG BLDPBD290069-1G
5-Bromo-1-methyl-4-nitro-1H-pyrazole 95%
5-Bromo-1-methyl-4-nitro-1H-pyrazole

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₄H₄BrN₃O₂
MW: 205,997 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD00195044
CAS-numero: 89607-13-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR