tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate 95%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD290087-50MGEA 158 EUR
BLDPBD290087-50MG BLDPBD290087-250MG BLDPBD290087-1G BLDPBD290087-100MG
tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate 95%
tert-Butyl (6-(hydroxymethyl)pyrimidin-4-yl)carbamate

Storage condition: Sealed in dry, 2-8°C

Kaava: C₁₀H₁₅N₃O₃
MW: 225,244 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD09952358
CAS-numero: 954097-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR