Bis(2,4-dimethylphenyl)amine 97%

BLDPBD297127-5GEA 184 EUR
BLDPBD297127-5G BLDPBD297127-250MG BLDPBD297127-10G BLDPBD297127-1G BLDPBD297127-0.025 BLDPBD297127-25G
Bis(2,4-dimethylphenyl)amine 97%
Bis(2,4-dimethylphenyl)amine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₁₆H₁₉N
MW: 225,34 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD16619146
CAS-numero: 19616-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR