1-(Methylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 96%

BLDPBD300643-100MGEA 117 EUR
BLDPBD300643-100MG BLDPBD300643-5G BLDPBD300643-250MG BLDPBD300643-1G
1-(Methylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 96%
1-(Methylsulphonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Storage condition: Sealed in dry, 2-8°C

Kaava: C₁₀H₁₇BN₂O₄S
MW: 272,129 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD18383275
CAS-numero: 944994-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR