1-Pyrenebutanoic acid Succinimidyl ester 98%

BLDPBD305485-1GEA 216 EUR
BLDPBD305485-1G BLDPBD305485-100MG BLDPBD305485-250MG
1-Pyrenebutanoic acid Succinimidyl ester 98%
1-Pyrenebutanoic acid Succinimidyl ester

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Kaava: C₂₄H₁₉NO₄
MW: 385,42 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00077403
CAS-numero: 114932-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR