3'-Sialyllactose sodium salt 90+% (HPAEC-PAD)

BLDPBD305537-250MGEA 182 EUR
BLDPBD305537-250MG BLDPBD305537-50MG
3'-Sialyllactose sodium salt 90+% (HPAEC-PAD)
3'-Sialyllactose sodium salt

Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C

Kaava: C₂₃H₃₈NNaO₁₉
MW: 655,53 g/mol
Storage Temperature: Pakastin
MDL numero: MFCD00166999
CAS-numero: 128596-80-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR