Diisopropyl phthalate 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD305884-25GEA 64.5 EUR
BLDPBD305884-25G BLDPBD305884-100G BLDPBD305884-5G
Diisopropyl phthalate 98%
Diisopropyl phthalate

Storage condition: Sealed in dry, Room Temperature

Kaava: C₁₄H₁₈O₄
MW: 250,2903 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00053717
CAS-numero: 605-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR