Tetrabutylammonium tetraphenylborate 98%
|
|
BLDPBD306176-25GEA
301
EUR
BLDPBD306176-25G
BLDPBD306176-5G
BLDPBD306176-1G
Tetrabutylammonium tetraphenylborate 98%
Tetrabutylammonium tetraphenylborate
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Kaava:
C₄₀H₅₆BN MW: 561,6904 g/mol Storage Temperature: Huoneenlämpö |
MDL numero:
MFCD00011749 CAS-numero: 15522-59-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...