4,4-Dimethyl-3-thiosemicarbazide 98%
|
|
BLDPBD306379-10GEA
254
EUR
BLDPBD306379-10G
BLDPBD306379-1G
BLDPBD306379-5G
4,4-Dimethyl-3-thiosemicarbazide 98%
4,4-Dimethyl-3-thiosemicarbazide
Storage condition: Keep in dark place, Inert atmosphere, Store in freezer, under -20°C
Kaava:
C₃H₉N₃S MW: 119,1887 g/mol Storage Temperature: Pakastin |
MDL numero:
MFCD00041308 CAS-numero: 6926-58-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...