2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane 98%

BLDPBD306408-25GEA 30.2 EUR
BLDPBD306408-25G BLDPBD306408-500G BLDPBD306408-100G
2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane 98%
2,2-Bis[4-(4-maleimidophenoxy)phenyl]propane

Storage condition: Sealed in dry, Room Temperature

Kaava: C₃₅H₂₆N₂O₆
MW: 570,60 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD09908251
CAS-numero: 79922-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR