2-Fluoro-5-formylphenylboronic acid pinacol ester 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD316401-100MGEA 40.6 EUR
BLDPBD316401-100MG BLDPBD316401-1G BLDPBD316401-250MG
2-Fluoro-5-formylphenylboronic acid pinacol ester 98%
2-Fluoro-5-formylphenylboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Kaava: C₁₃H₁₆BFO₃
MW: 250,07 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD11867877
CAS-numero: 1112208-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR