1,4-Dichloroisoquinoline 97%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD32058-10GEA 295 EUR
BLDPBD32058-10G BLDPBD32058-250MG BLDPBD32058-5G BLDPBD32058-1G
1,4-Dichloroisoquinoline 97%
1,4-Dichloroisoquinoline

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₉H₅Cl₂N
MW: 198,0500 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD05664199
CAS-numero: 15298-58-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR