Thiophene-3,4-dicarbonitrile 97%

BLDPBD32412-5GEA 206 EUR
BLDPBD32412-5G BLDPBD32412-250MG BLDPBD32412-25G BLDPBD32412-1G BLDPBD32412-10G
Thiophene-3,4-dicarbonitrile 97%
Thiophene-3,4-dicarbonitrile

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Kaava: C₆H₂N₂S
MW: 134,158 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD01233047
CAS-numero: 18853-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR