4-Fluoro-3-nitrophenol 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD3280-25GEA 256 EUR
BLDPBD3280-0.025G BLDPBD3280-25G BLDPBD3280-1G BLDPBD3280-5G BLDPBD3280-10G BLDPBD3280-250MG
4-Fluoro-3-nitrophenol 98%
4-Fluoro-3-nitrophenol

Storage condition: Inert atmosphere, Room Temperature

Kaava: C₆H₄FNO₃
MW: 157,0993 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD04038714
CAS-numero: 2105-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR