Thieno[2,3-d]pyrimidin-4(3H)-one 98%

BLDPBD3293-250MGEA 7.3 EUR
BLDPBD3293-250MG BLDPBD3293-1G BLDPBD3293-5G BLDPBD3293-100G BLDPBD3293-25G BLDPBD3293-10G
Thieno[2,3-d]pyrimidin-4(3H)-one 98%
Thieno[2,3-d]pyrimidin-4(3H)-one

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₆H₄N₂OS
MW: 152,1738 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00706456
CAS-numero: 14080-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR