3-Amino-2-bromo-5-methylpyridine 98%

BLDPBD33317-25GEA 226 EUR
BLDPBD33317-25G BLDPBD33317-1G BLDPBD33317-0.025G BLDPBD33317-5G
3-Amino-2-bromo-5-methylpyridine 98%
3-Amino-2-bromo-5-methylpyridine

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Kaava: C₆H₇BrN₂
MW: 187,04 g/mol
Storage Temperature: Jääkaappi
MDL numero: MFCD08277281
CAS-numero: 34552-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR