D-(+)-Cystine 98%

BLDPBD33339-1GEA 7.3 EUR
BLDPBD33339-1G BLDPBD33339-500G BLDPBD33339-25G BLDPBD33339-250MG BLDPBD33339-100G BLDPBD33339-5G
D-(+)-Cystine 98%
D-(+)-Cystine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Kaava: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240,3 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002610
CAS-numero: 349-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR