4-Fluoro-N-methylbenzylamine 98%

Toimittaja: BLD PHARMATECH GMBH

BLDPBD33605-25GEA 264 EUR
BLDPBD33605-25G BLDPBD33605-1G BLDPBD33605-5G
4-Fluoro-N-methylbenzylamine 98%
4-Fluoro-N-methylbenzylamine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Kaava: C₈H₁₀FN
MW: 139,17 g/mol
Storage Temperature: Huoneenlämpö
MDL numero: MFCD03839848
CAS-numero: 405-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR